missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha,Nepsilon-Bis(tert-butoxycarbonyl)-L-lysine Dicyclohexylammonium Salt 98.0+%, TCI America™
$33.56 - $106.25
Chemical Identifiers
| CAS | 15098-69-8 |
|---|---|
| Molecular Formula | C28H53N3O6 |
| Molecular Weight (g/mol) | 527.75 |
| MDL Number | MFCD00038892 |
| InChI Key | HRLHJTYAMCGERD-VAMKTSTMNA-N |
| Synonym | boc-lys boc-oh.dcha, boc-lys boc-oh dcha, dicyclohexylamine s-2,6-bis tert-butoxycarbonyl amino hexanoate, n,n'-di-boc-l-lysine dicyclohexylamine salt, boc-lys boc-oh dicyclohexammonium salt, n,n'-bis tert-butoxycarbonyl-l-lysine dicyclohexylamine 1:1, 2s-2,6-bis tert-butoxycarbonyl amino hexanoic acid; dicha, boc-lys boc-ohcha, boc-lys boc-oh?cha, boc-lys boc-ohdcha |
| PubChem CID | 12017189 |
| IUPAC Name | (2S)-2,6-bis({[(tert-butoxy)carbonyl]amino})hexanoic acid; N-cyclohexylcyclohexanamine |
| SMILES | C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O |
Chemical Identifiers
| 15098-69-8 | |
| 527.75 | |
| HRLHJTYAMCGERD-VAMKTSTMNA-N | |
| 12017189 | |
| C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O |
| C28H53N3O6 | |
| MFCD00038892 | |
| boc-lys boc-oh.dcha, boc-lys boc-oh dcha, dicyclohexylamine s-2,6-bis tert-butoxycarbonyl amino hexanoate, n,n'-di-boc-l-lysine dicyclohexylamine salt, boc-lys boc-oh dicyclohexammonium salt, n,n'-bis tert-butoxycarbonyl-l-lysine dicyclohexylamine 1:1, 2s-2,6-bis tert-butoxycarbonyl amino hexanoic acid; dicha, boc-lys boc-ohcha, boc-lys boc-oh?cha, boc-lys boc-ohdcha | |
| (2S)-2,6-bis({[(tert-butoxy)carbonyl]amino})hexanoic acid; N-cyclohexylcyclohexanamine |
Specifications
| 15098-69-8 | |
| White | |
| MFCD00038892 | |
| boc-lys boc-oh.dcha, boc-lys boc-oh dcha, dicyclohexylamine s-2,6-bis tert-butoxycarbonyl amino hexanoate, n,n'-di-boc-l-lysine dicyclohexylamine salt, boc-lys boc-oh dicyclohexammonium salt, n,n'-bis tert-butoxycarbonyl-l-lysine dicyclohexylamine 1:1, 2s-2,6-bis tert-butoxycarbonyl amino hexanoic acid; dicha, boc-lys boc-ohcha, boc-lys boc-oh?cha, boc-lys boc-ohdcha | |
| C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O | |
| 527.75 | |
| 527.75 | |
| Crystalline Powder |
| 141°C | |
| C28H53N3O6 | |
| 5 g | |
| HRLHJTYAMCGERD-VAMKTSTMNA-N | |
| (2S)-2,6-bis({[(tert-butoxy)carbonyl]amino})hexanoic acid; N-cyclohexylcyclohexanamine | |
| 12017189 | |
| ≥98.0% (HPLC,T) | |
| Nalpha,Nepsilon-Bis(tert-butoxycarbonyl)-L-lysine Dicyclohexylammonium Salt |
Safety and Handling
TSCA : No