accessibility menu, dialog, popup

UserName

CAS RN 15098-69-8

H3CH3CCH3OONH(S)OHONHOOCH3CH3H3CHN

CAS RN 15098-69-8

IUPAC Name: (2S)-2,6-bis({[(tert-butoxy)carbonyl]amino})hexanoic acid; N-cyclohexylcyclohexanamine
Synonyms: (2S)-2,6-bis[(tert-butoxycarbonyl)amino]hexanoic acid; dicha
Molecular Weight (g/mol): 527.75
Molecular Formula: C28H53N3O6
InChi Key: HRLHJTYAMCGERD-VAMKTSTMNA-N
SMILES: C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)