missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha,Nepsilon-Bis(tert-butoxycarbonyl)-L-lysine Dicyclohexylammonium Salt 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 141°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 527.75 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Nalpha,Nepsilon-Bis(tert-butoxycarbonyl)-L-lysine Dicyclohexylammonium Salt |
Safety and Handling
| TSCA | No |