missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Fmoc-S-trityl-D-cysteine, 98%

Catalog No. p-7041488
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6230103 1 g
AAH6230106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6230103 Supplier Thermo Scientific Chemicals Supplier No. H6230103
Only null left

CAS: 167015-11-4 | C37H31NO4S | 585.718 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 167015-11-4
Molecular Formula C37H31NO4S
Molecular Weight (g/mol) 585.718
MDL Number MFCD00151922
InChI Key KLBPUVPNPAJWHZ-UUWRZZSWSA-N
Synonym fmoc-d-cys trt-oh, n-fmoc-s-trityl-d-cysteine, fmoc-s-trityl-d-cysteine, n-alpha-9-fluorenylmethyloxycarbonyl-s-trityl-d-cysteine, n-9h-fluoren-9-ylmethoxy carbonyl-s-triphenylmethyl-d-cysteine, d-cysteine, n-9h-fluoren-9-ylmethoxy carbonyl-s-triphenylmethyl, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-triphenylmethyl sulfanyl propanoic acid, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tritylthio propanoic acid, ambotzfaa1035, pubchem19012
PubChem CID 7168037
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-tritylsulfanylpropanoic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)SCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46

Specifications

Melting Point 172°C to 176°C
Quantity 1 g
Beilstein 7558914
Formula Weight 587.71
Percent Purity 98%
Chemical Name or Material Nalpha-Fmoc-S-trityl-D-cysteine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only