missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Fmoc-O-tert-butyl-L-threonine, 98%

Catalog No. AAH5980809
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5980809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5980809 Supplier Thermo Scientific Chemicals Supplier No. H5980809
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 71989-35-0 | C23H27NO5 | 397.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71989-35-0
Molecular Formula C23H27NO5
Molecular Weight (g/mol) 397.47
MDL Number MFCD00077075
InChI Key LZOLWEQBVPVDPR-VLIAUNLRSA-N
Synonym fmoc-thr tbu-oh, fmoc-o-tert-butyl-l-threonine, fmoc-thr t-bu-oh, n-9-fluorenylmethoxycarbonyl-o-tert-butyl-l-threonine, fmoc-thr but, 2s,3r-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, n-9h-fluoren-9-yl methoxy carbonyl-o-tert-butyl-l-threonine, fmoc-thr but-oh, fmoc-o-tert.butyl-l-threonine, fmoc-thr otbu-oh
PubChem CID 6364643
IUPAC Name (2S,3R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]butanoic acid
SMILES C[C@@H](OC(C)(C)C)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

Specifications

Melting Point 131°C to 134°C
Quantity 10 g
Beilstein 4581133
Optical Rotation +16° (c=1 in Ethyl Acetate)
Formula Weight 397.46
Percent Purity 98%
Chemical Name or Material Nalpha-Fmoc-O-tert-butyl-L-threonine
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 276-261-1
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only