missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Fmoc-Nepsilon-benzyloxycarbonyl-L-lysine, 98%

Catalog No. p-7041625
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6286606 5 g
AAH6286614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6286606 Supplier Thermo Scientific Chemicals Supplier No. H6286606
Only null left

CAS: 86060-82-4 | C29H30N2O6 | 502.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 86060-82-4
Molecular Formula C29H30N2O6
Molecular Weight (g/mol) 502.57
MDL Number MFCD00065662
InChI Key KRULQRVJXQQPQH-CYLJNIGPNA-N
Synonym fmoc-lys z-oh, fmoc-lys cbz-oh, n, a-fmoc-n, a-z-l-lysine, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-6-phenylmethoxycarbonylamino hexanoic acid, 2s-6-benzyloxy carbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, lysine derivative 4, pubchem19681, fmoc-l-lys z-oh, fmoc-n-epsilon-cbz-l-lysine, n alpha-fmoc-n epsilon-z-l-lysine
PubChem CID 3013736
SMILES OC(=O)[C@H](CCCCNC(=O)OCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 110°C to 120°C
Quantity 5 g
Beilstein 3578530
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-(phenylmethoxycarbonylamino)hexanoic acid
Formula Weight 502.56
Percent Purity 98%
Chemical Name or Material Nalpha-Fmoc-Nepsilon-benzyloxycarbonyl-L-lysine
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only