missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-Fmoc-Ndelta-trityl-L-glutamine, 98%
CAS: 132327-80-1 | C39H34N2O5 | 610.71 g/mol
$120.21 - $437.79
Chemical Identifiers
| CAS | 132327-80-1 |
|---|---|
| Molecular Formula | C39H34N2O5 |
| Molecular Weight (g/mol) | 610.71 |
| MDL Number | MFCD00077056 |
| InChI Key | WDGICUODAOGOMO-DHUJRADRSA-N |
| Synonym | fmoc-gln trt-oh, fmoc-l-gln trt-oh, fmoc-glutamine trt-oh, n-fmoc-n5-trityl-l-glutamine, fmoc-glutamine trt-oh, a-fmoc-n, a-trityl-l-glutamine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-oxo-5-tritylamino pentanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-triphenylmethylcarbamoyl butanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-oxo-5-tritylamino pentanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-triphenylmethylcarbamoyl butanoic acid, pubchem10017 |
| PubChem CID | 10919157 |
| IUPAC Name | (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-(tritylamino)pentanoic acid |
| SMILES | C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)NC(=O)CCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5908006
|
Thermo Scientific Chemicals
H5908006 |
5 g |
Each for $120.21
|
|
|||||
|
AAH5908014
|
Thermo Scientific Chemicals
H5908014 |
25 g |
Each for $437.79
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 132327-80-1 | |
| 610.71 | |
| WDGICUODAOGOMO-DHUJRADRSA-N | |
| 10919157 | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)NC(=O)CCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46 |
| C39H34N2O5 | |
| MFCD00077056 | |
| fmoc-gln trt-oh, fmoc-l-gln trt-oh, fmoc-glutamine trt-oh, n-fmoc-n5-trityl-l-glutamine, fmoc-glutamine trt-oh, a-fmoc-n, a-trityl-l-glutamine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-oxo-5-tritylamino pentanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-triphenylmethylcarbamoyl butanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-oxo-5-tritylamino pentanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-triphenylmethylcarbamoyl butanoic acid, pubchem10017 | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-(tritylamino)pentanoic acid |
Specifications
| 132327-80-1 | |
| C39H34N2O5 | |
| 5 g | |
| Moisture sensitive | |
| WDGICUODAOGOMO-DHUJRADRSA-N | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)NC(=O)CCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46 | |
| 610.71 | |
| 610.71 | |
| Nalpha-Fmoc-Ndelta-trityl-L-glutamine |
| 165°C to 172°C | |
| MFCD00077056 | |
| 4343953 | |
| fmoc-gln trt-oh, fmoc-l-gln trt-oh, ksc180c8t, n-fmoc-n5-trityl-l-glutamine, fmoc-glutamine trt-oh, a-fmoc-n, a-trityl-l-glutamine, a-fmoc-n, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-triphenylmethylcarbamoyl butanoic acid, pubchem10017, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-triphenylmethylcarbamoyl butanoic acid, pubchem10017 | |
| −14° (c=1 in DMF) | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-(tritylamino)pentanoic acid | |
| 10919157 | |
| 98% |
Safety and Handling
GHS H Statement
H317
May cause an allergic skin reaction.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only