missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Fmoc-Ndelta-trityl-L-glutamine, 98%

Catalog No. AAH5908006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5908006 5 g
AAH5908014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5908006 Supplier Thermo Scientific Chemicals Supplier No. H5908006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 132327-80-1 | C39H34N2O5 | 610.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 132327-80-1
Molecular Formula C39H34N2O5
Molecular Weight (g/mol) 610.71
MDL Number MFCD00077056
InChI Key WDGICUODAOGOMO-DHUJRADRSA-N
Synonym fmoc-gln trt-oh, nalpha-fmoc-ndelta-trityl-l-glutamine, fmoc-glutamine trt-oh, n-fmoc-n5-trityl-l-glutamine, n, a-fmoc-n, a-trityl-l-glutamine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-oxo-5-tritylamino pentanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-triphenylmethylcarbamoyl butanoic acid, pubchem10017, fmoc-l-gln trt-oh, ksc180c8t
PubChem CID 10919157
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-(tritylamino)pentanoic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)NC(=O)CCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46

Specifications

Melting Point 165°C to 172°C
Quantity 5 g
Beilstein 4343953
Sensitivity Moisture sensitive
Optical Rotation −14° (c=1 in DMF)
Formula Weight 610.71
Percent Purity 98%
Chemical Name or Material Nalpha-Fmoc-Ndelta-trityl-L-glutamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only