missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-Fmoc-N^d-trityl-D-glutamine, 98%, Thermo Scientific Chemicals
$294.81 - $294.81
Chemical Identifiers
| CAS | 200623-62-7 |
|---|---|
| Molecular Formula | C39H34N2O5 |
| Molecular Weight (g/mol) | 610.71 |
| MDL Number | MFCD00151924 |
| InChI Key | WDGICUODAOGOMO-PGUFJCEWSA-N |
| Synonym | fmoc-d-gln trt-oh, n-fmoc-n'-trityl-d-glutamine, fmoc-gln trt, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-triphenylmethylcarbamoyl butanoic acid, ambotzfaa1322, pubchem12399, n-alpha-9-fluorenylmethyloxycarbonyl-n-gamma-trityl-d-glutamine, n∼2∼-9h-fluoren-9-yl methoxy carbonyl-n-triphenylmethyl-d-glutamine, 2r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-triphenylmethyl carbamoyl butanoic acid |
| PubChem CID | 24820181 |
| IUPAC Name | (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-(tritylamino)pentanoic acid |
| SMILES | C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)NC(=O)CCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6253906
|
Thermo Scientific Chemicals
H6253906 |
5 g |
Each for $294.81
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 200623-62-7 | |
| 610.71 | |
| WDGICUODAOGOMO-PGUFJCEWSA-N | |
| 24820181 | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)NC(=O)CCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46 |
| C39H34N2O5 | |
| MFCD00151924 | |
| fmoc-d-gln trt-oh, n-fmoc-n'-trityl-d-glutamine, fmoc-gln trt, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-triphenylmethylcarbamoyl butanoic acid, ambotzfaa1322, pubchem12399, n-alpha-9-fluorenylmethyloxycarbonyl-n-gamma-trityl-d-glutamine, n∼2∼-9h-fluoren-9-yl methoxy carbonyl-n-triphenylmethyl-d-glutamine, 2r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-triphenylmethyl carbamoyl butanoic acid | |
| (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-(tritylamino)pentanoic acid |
Specifications
| 200623-62-7 | |
| MFCD00151924 | |
| fmoc-d-gln trt-oh, n-fmoc-n'-trityl-d-glutamine, fmoc-gln trt, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-triphenylmethylcarbamoyl butanoic acid, ambotzfaa1322, pubchem12399, n-alpha-9-fluorenylmethyloxycarbonyl-n-gamma-trityl-d-glutamine, n∼2∼-9h-fluoren-9-yl methoxy carbonyl-n-triphenylmethyl-d-glutamine, 2r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-triphenylmethyl carbamoyl butanoic acid | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)NC(=O)CCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46 | |
| 610.71 | |
| 610.7 | |
| Nalpha-Fmoc-Ndelta-trityl-D-glutamine |
| C39H34N2O5 | |
| 5 g | |
| WDGICUODAOGOMO-PGUFJCEWSA-N | |
| (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-(tritylamino)pentanoic acid | |
| 24820181 | |
| 98% |
Safety and Handling
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only