missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-Fmoc-N^w-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulfonyl)-L-arginine, 98%
CAS: 154445-77-9 | C34H40N4O7S | 648.775 g/mol
$134.52 - $570.19
Chemical Identifiers
| CAS | 154445-77-9 |
|---|---|
| Molecular Formula | C34H40N4O7S |
| Molecular Weight (g/mol) | 648.775 |
| MDL Number | MFCD00235804 |
| InChI Key | HNICLNKVURBTKV-NDEPHWFRSA-N |
| Synonym | fmoc-arg pbf-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-3-2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl sulfonyl guanidino pentanoic acid, nalpha-fmoc-nomega-pbf-l-arginine, fmoc-arg pbf, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-n'-2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-ylsulfonyl carbamimidamido pentanoic acid, fmoc-arg pdf-oh, pubchem9946, fmoc-l-arg pbf-oh, fmoc-arg pbf-oh hplc, n-alpha-9-fluorenylmethoxycarbonyl-n-omega-2,2,4 |
| PubChem CID | 11354259 |
| IUPAC Name | (2S)-5-[[amino-[(2,2,4,6,7-pentamethyl-3H-1-benzofuran-5-yl)sulfonylamino]methylidene]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid |
| SMILES | CC1=C2C(=C(C(=C1C)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)N)C)CC(O2)(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5903906
|
Thermo Scientific Chemicals
H5903906 |
5 g |
Each for $134.52
|
|
|||||
|
AAH5903914
|
Thermo Scientific Chemicals
H5903914 |
25 g |
Each for $570.19
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 154445-77-9 | |
| 648.775 | |
| HNICLNKVURBTKV-NDEPHWFRSA-N | |
| 11354259 | |
| CC1=C2C(=C(C(=C1C)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)N)C)CC(O2)(C)C |
| C34H40N4O7S | |
| MFCD00235804 | |
| fmoc-arg pbf-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-3-2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl sulfonyl guanidino pentanoic acid, nalpha-fmoc-nomega-pbf-l-arginine, fmoc-arg pbf, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-n'-2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-ylsulfonyl carbamimidamido pentanoic acid, fmoc-arg pdf-oh, pubchem9946, fmoc-l-arg pbf-oh, fmoc-arg pbf-oh hplc, n-alpha-9-fluorenylmethoxycarbonyl-n-omega-2,2,4 | |
| (2S)-5-[[amino-[(2,2,4,6,7-pentamethyl-3H-1-benzofuran-5-yl)sulfonylamino]methylidene]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid |
Specifications
| 154445-77-9 | |
| MFCD00235804 | |
| 8302671 | |
| HNICLNKVURBTKV-NDEPHWFRSA-N | |
| CC1=C2C(=C(C(=C1C)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)N)C)CC(O2)(C)C | |
| 648.775 | |
| 648.77 | |
| Nalpha-Fmoc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulfonyl)-L-arginine |
| C34H40N4O7S | |
| 5 g | |
| fmoc-arg pbf-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-3-2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl sulfonyl guanidino pentanoic acid, nalpha-fmoc-nomega-pbf-l-arginine, fmoc-arg pbf, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-n'-2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-ylsulfonyl carbamimidamido pentanoic acid, fmoc-arg pdf-oh, pubchem9946, fmoc-l-arg pbf-oh, fmoc-arg pbf-oh hplc, n-alpha-9-fluorenylmethoxycarbonyl-n-omega-2,2,4 | |
| −5.5° (c=1 in DMF) | |
| (2S)-5-[[amino-[(2,2,4,6,7-pentamethyl-3H-1-benzofuran-5-yl)sulfonylamino]methylidene]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid | |
| 11354259 | |
| 98% |
Safety and Handling
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only