missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Fmoc-N^w-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulfonyl)-L-arginine, 98%

Catalog No. AAH5903906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5903906 5 g
AAH5903914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5903906 Supplier Thermo Scientific Chemicals Supplier No. H5903906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 154445-77-9 | C34H40N4O7S | 648.775 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 154445-77-9
Molecular Formula C34H40N4O7S
Molecular Weight (g/mol) 648.775
MDL Number MFCD00235804
InChI Key HNICLNKVURBTKV-NDEPHWFRSA-N
Synonym fmoc-arg pbf-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-3-2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl sulfonyl guanidino pentanoic acid, nalpha-fmoc-nomega-pbf-l-arginine, fmoc-arg pbf, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-n'-2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-ylsulfonyl carbamimidamido pentanoic acid, fmoc-arg pdf-oh, pubchem9946, fmoc-l-arg pbf-oh, fmoc-arg pbf-oh hplc, n-alpha-9-fluorenylmethoxycarbonyl-n-omega-2,2,4
PubChem CID 11354259
IUPAC Name (2S)-5-[[amino-[(2,2,4,6,7-pentamethyl-3H-1-benzofuran-5-yl)sulfonylamino]methylidene]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
SMILES CC1=C2C(=C(C(=C1C)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)N)C)CC(O2)(C)C

Specifications

Quantity 5 g
Beilstein 8302671
Optical Rotation −5.5° (c=1 in DMF)
Formula Weight 648.77
Percent Purity 98%
Chemical Name or Material Nalpha-Fmoc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulfonyl)-L-arginine
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only