missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Fmoc-L-lysine hydrochloride, 98%

Catalog No. p-7040077
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5969603 1 g
AAH5969606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5969603 Supplier Thermo Scientific Chemicals Supplier No. H5969603
Only null left

CAS: 139262-23-0 | C21H25ClN2O4 | 404.89 g/mol

It is used as an active pharmaceutical intermediate and in the synthesis of nε-(7-diethylaminocoumarin-3-carboxyl)- and nε-(7-methoxycoumarin-3-carboxyl)-l-fmoc lysine as tools for protease cleavage detection by fluorescence.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate and in the synthesis of nε-(7-diethylaminocoumarin-3-carboxyl)- and nε-(7-methoxycoumarin-3-carboxyl)-l-fmoc lysine as tools for protease cleavage detection by fluorescence.

Solubility
Soluble in dimethyl formamide (0.3g in 2ml).

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 139262-23-0
Molecular Formula C21H25ClN2O4
Molecular Weight (g/mol) 404.89
MDL Number MFCD00190889
InChI Key MVMZFAIUUXYFGY-UPGKADEYNA-N
Synonym fmoc-lys-oh.hcl, fmoc-lys-oh hydrochloride, fmoc-lys-oh hcl, nalpha-fmoc-l-lysine hydrochloride, fmoc-l-lysine hydrochloride, nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-lysine hydrochloride, l-lysine, n2-9h-fluoren-9-ylmethoxy carbonyl-, monohydrochloride, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-aminohexanoic acid hydrochloride, fmoc-l-lysine hcl, fmoc-lys-oh. hcl
PubChem CID 11858928
IUPAC Name (2S)-6-amino-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid;hydrochloride
SMILES Cl.NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Quantity 1 g
Beilstein 8663370
Solubility Information Soluble in dimethyl formamide (0.3g in 2ml).
Formula Weight 404.89
Percent Purity 98%
Chemical Name or Material Nalpha-Fmoc-L-lysine hydrochloride
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only