missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-Fmoc-L-arginine, 95%
CAS: 91000-69-0 | C21H24N4O4 | 396.45 g/mol
$95.40 - $362.12
Chemical Identifiers
| CAS | 91000-69-0 |
|---|---|
| Molecular Formula | C21H24N4O4 |
| Molecular Weight (g/mol) | 396.45 |
| MDL Number | MFCD00051770 |
| InChI Key | DVBUCBXGDWWXNY-SFHVURJKSA-N |
| Synonym | fmoc-arg-oh, fmoc-l-arginine, nalpha-fmoc-l-arginine, alfa-fmoc-l-arginine, fmoc-l-arg-oh, n-a-fmoc-l-arginine, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-5-guanidino-pentanoic acid, 2s-5-amidinoamino-2-fluoren-9-ylmethoxy carbonylamino pentanoic acid, 2s-5-carbamimidamido-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, fmoc-arg |
| PubChem CID | 2724631 |
| IUPAC Name | (2S)-5-{[amino(iminiumyl)methyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoate |
| SMILES | NC(=[NH2+])NCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6666506
|
Thermo Scientific Chemicals
H6666506 |
5 g |
Each for $95.40
|
|
|||||
|
AAH6666514
|
Thermo Scientific Chemicals
H6666514 |
25 g |
Each for $362.12
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 91000-69-0 | |
| 396.45 | |
| DVBUCBXGDWWXNY-SFHVURJKSA-N | |
| 2724631 | |
| NC(=[NH2+])NCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O |
| C21H24N4O4 | |
| MFCD00051770 | |
| fmoc-arg-oh, fmoc-l-arginine, nalpha-fmoc-l-arginine, alfa-fmoc-l-arginine, fmoc-l-arg-oh, n-a-fmoc-l-arginine, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-5-guanidino-pentanoic acid, 2s-5-amidinoamino-2-fluoren-9-ylmethoxy carbonylamino pentanoic acid, 2s-5-carbamimidamido-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, fmoc-arg | |
| (2S)-5-{[amino(iminiumyl)methyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoate |
Specifications
| 91000-69-0 | |
| C21H24N4O4 | |
| 4828015 | |
| DVBUCBXGDWWXNY-SFHVURJKSA-N | |
| (2S)-5-{[amino(iminiumyl)methyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoate | |
| 396.45 | |
| 396.45 | |
| 95% | |
| Nalpha-Fmoc-L-arginine |
| White | |
| MFCD00051770 | |
| fmoc-arg-oh, fmoc-l-arginine, nalpha-fmoc-l-arginine, alfa-fmoc-l-arginine, fmoc-l-arg-oh, n-a-fmoc-l-arginine, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-5-guanidino-pentanoic acid, 2s-5-amidinoamino-2-fluoren-9-ylmethoxy carbonylamino pentanoic acid, 2s-5-carbamimidamido-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, fmoc-arg | |
| NC(=[NH2+])NCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O | |
| 5 g | |
| 2724631 | |
| Moisture sensitive | |
| Powder |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only