missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-Carbobenzoxy-L-arginine 97.0+%, TCI America™
$65.24 - $173.37
Chemical Identifiers
| CAS | 1234-35-1 |
|---|---|
| Molecular Formula | C14H20N4O4 |
| Molecular Weight (g/mol) | 308.338 |
| MDL Number | MFCD00001762 |
| InChI Key | SJSSFUMSAFMFNM-NSHDSACASA-N |
| Synonym | z-arg-oh, nalpha-cbz-l-arginine, nalpha-carbobenzyloxy-l-arginine, cbz-arg-oh, z-l-arg-oh, cbz-l-arginine, cbz-l-arg-oh, n-benzyloxycarbonyl-l-arginine, carbobenzoxy-l-arginine, z-l-arginine |
| PubChem CID | 71055 |
| IUPAC Name | (2S)-5-(diaminomethylideneamino)-2-(phenylmethoxycarbonylamino)pentanoic acid |
| SMILES | C1=CC=C(C=C1)COC(=O)NC(CCCN=C(N)N)C(=O)O |
Chemical Identifiers
| 1234-35-1 | |
| 308.338 | |
| SJSSFUMSAFMFNM-NSHDSACASA-N | |
| 71055 | |
| C1=CC=C(C=C1)COC(=O)NC(CCCN=C(N)N)C(=O)O |
| C14H20N4O4 | |
| MFCD00001762 | |
| z-arg-oh, nalpha-cbz-l-arginine, nalpha-carbobenzyloxy-l-arginine, cbz-arg-oh, z-l-arg-oh, cbz-l-arginine, cbz-l-arg-oh, n-benzyloxycarbonyl-l-arginine, carbobenzoxy-l-arginine, z-l-arginine | |
| (2S)-5-(diaminomethylideneamino)-2-(phenylmethoxycarbonylamino)pentanoic acid |
Specifications
| 1234-35-1 | |
| White | |
| MFCD00001762 | |
| z-arg-oh, nalpha-cbz-l-arginine, nalpha-carbobenzyloxy-l-arginine, cbz-arg-oh, z-l-arg-oh, cbz-l-arginine, cbz-l-arg-oh, n-benzyloxycarbonyl-l-arginine, carbobenzoxy-l-arginine, z-l-arginine | |
| C1=CC=C(C=C1)COC(=O)NC(CCCN=C(N)N)C(=O)O | |
| 308.338 | |
| 308.34 | |
| Crystalline Powder |
| 173°C | |
| C14H20N4O4 | |
| 1 g | |
| SJSSFUMSAFMFNM-NSHDSACASA-N | |
| (2S)-5-(diaminomethylideneamino)-2-(phenylmethoxycarbonylamino)pentanoic acid | |
| 71055 | |
| ≥97.0% (T) | |
| Nalpha-Carbobenzoxy-L-arginine |
Safety and Handling
TSCA : Yes