CAS RN 1234-35-1
CAS RN 1234-35-1
IUPAC Name:
(2S)-5-{[amino(iminiumyl)methyl]amino}-2-{[(benzyloxy)carbonyl]amino}pentanoate
Synonyms:
(2S)-5-{[amino(iminio)methyl]amino}-2-{[(benzyloxy)carbonyl]amino}pentanoate
Molecular Weight (g/mol):
308.34
Molecular Formula:
C14H20N4O4
InChi Key:
SJSSFUMSAFMFNM-NSHDSACASA-N
SMILES:
NC(=[NH2+])NCCC[C@H](NC(=O)OCC1=CC=CC=C1)C([O-])=O
1
–
1
of
1
results
1
Nalpha-Carbobenzoxy-L-arginine 97.0+%, TCI America™
CAS: 1234-35-1 Molecular Formula: C14H20N4O4 Molecular Weight (g/mol): 308.338 MDL Number: MFCD00001762 InChI Key: SJSSFUMSAFMFNM-NSHDSACASA-N Synonym: z-arg-oh,nalpha-cbz-l-arginine,nalpha-carbobenzyloxy-l-arginine,cbz-arg-oh,z-l-arg-oh,cbz-l-arginine,cbz-l-arg-oh,n-benzyloxycarbonyl-l-arginine,carbobenzoxy-l-arginine,z-l-arginine PubChem CID: 71055 IUPAC Name: (2S)-5-(diaminomethylideneamino)-2-(phenylmethoxycarbonylamino)pentanoic acid SMILES: C1=CC=C(C=C1)COC(=O)NC(CCCN=C(N)N)C(=O)O