missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-Carbobenzoxy-D-histidine 98.0+%, TCI America™
$108.11 - $318.75
Chemical Identifiers
| CAS | 67424-93-5 |
|---|---|
| Molecular Formula | C14H15N3O4 |
| Molecular Weight (g/mol) | 289.291 |
| MDL Number | MFCD00067702 |
| InChI Key | WCOJOHPAKJFUDF-GFCCVEGCSA-N |
| Synonym | z-d-his-oh, nalpha-cbz-d-histidine, cbz-d-his-oh, cbz-d-histidine, z-d-histidine, nalpha-carbobenzoxy-d-histidine, n alpha-cbz-d-histidine, n alpha-benzyloxycarbonyl-d-histidine, n-benzyloxy carbonyl-d-histidine, d-histidine,n-phenylmethoxy carbonyl |
| PubChem CID | 736161 |
| IUPAC Name | (2R)-3-(1H-imidazol-5-yl)-2-(phenylmethoxycarbonylamino)propanoic acid |
| SMILES | C1=CC=C(C=C1)COC(=O)NC(CC2=CN=CN2)C(=O)O |
Chemical Identifiers
| 67424-93-5 | |
| 289.291 | |
| WCOJOHPAKJFUDF-GFCCVEGCSA-N | |
| 736161 | |
| C1=CC=C(C=C1)COC(=O)NC(CC2=CN=CN2)C(=O)O |
| C14H15N3O4 | |
| MFCD00067702 | |
| z-d-his-oh, nalpha-cbz-d-histidine, cbz-d-his-oh, cbz-d-histidine, z-d-histidine, nalpha-carbobenzoxy-d-histidine, n alpha-cbz-d-histidine, n alpha-benzyloxycarbonyl-d-histidine, n-benzyloxy carbonyl-d-histidine, d-histidine,n-phenylmethoxy carbonyl | |
| (2R)-3-(1H-imidazol-5-yl)-2-(phenylmethoxycarbonylamino)propanoic acid |
Specifications
| 67424-93-5 | |
| C14H15N3O4 | |
| 1 g | |
| WCOJOHPAKJFUDF-GFCCVEGCSA-N | |
| (2R)-3-(1H-imidazol-5-yl)-2-(phenylmethoxycarbonylamino)propanoic acid | |
| 736161 | |
| ≥98.0% (T) | |
| Nalpha-Carbobenzoxy-D-histidine |
| 163°C | |
| MFCD00067702 | |
| z-d-his-oh, nalpha-cbz-d-histidine, cbz-d-his-oh, cbz-d-histidine, z-d-histidine, nalpha-carbobenzoxy-d-histidine, n alpha-cbz-d-histidine, n alpha-benzyloxycarbonyl-d-histidine, n-benzyloxy carbonyl-d-histidine, d-histidine,n-phenylmethoxy carbonyl | |
| C1=CC=C(C=C1)COC(=O)NC(CC2=CN=CN2)C(=O)O | |
| 289.291 | |
| 289.29 | |
| Crystalline Powder |
Safety and Handling
TSCA : No