missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-Nepsilon-Fmoc-D-lysine, 95%

Catalog No. AAH6211606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6211606 5 g
AAH6211603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6211606 Supplier Thermo Scientific Chemicals Supplier No. H6211606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 115186-31-7 | C26H32N2O6 | 468.55 g/mol

Nalpha-Boc-Nepsilon-Fmoc-D-lysine is commonly used in the synthesis of aza-amino acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nalpha-Boc-Nepsilon-Fmoc-D-lysine is commonly used in the synthesis of aza-amino acids.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 115186-31-7
Molecular Formula C26H32N2O6
Molecular Weight (g/mol) 468.55
MDL Number MFCD00076966
InChI Key JYEVQYFWINBXJU-ANBDAQEENA-N
Synonym boc-d-lys fmoc-oh, n-boc-n'-fmoc-d-lysine, boc-l-lys fmoc-oh, boc-n-epsilon-fmoc-d-lysine, n-alpha-t-butyloxycarbonyl-n-epsilon-9-fluorenylmethyloxycarbonyl-d-lysine, 2r-2-tert-butoxycarbonyl amino-6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, ambotzbaa1040, n-boc-n-fmoc-d-lysine, pubchem12013, n2-boc-n6-fmoc-d-lysine
PubChem CID 14019424
IUPAC Name (2R)-6-(9H-fluoren-9-ylmethoxycarbonylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 117°C to 123°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 468.55
Percent Purity 95%
Chemical Name or Material Nalpha-Boc-Nepsilon-Fmoc-D-lysine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only