missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-N^w-nitro-L-arginine, 98%

CAS: 2188-18-3 | C11H21N5O6 | 319.318 g/mol

$164.36 - $508.36

Chemical Identifiers

CAS 2188-18-3
Molecular Formula C11H21N5O6
Molecular Weight (g/mol) 319.318
MDL Number MFCD00065556
InChI Key OZSSOVRIEPAIMP-ZETCQYMHSA-N
Synonym boc-arg no2-oh, s-2-tert-butoxycarbonyl amino-5-3-nitroguanidino pentanoic acid, n2-tert-butoxycarbonyl-n5-n-nitrocarbamimidoyl-l-ornithine, 2s-2-tert-butoxycarbonyl amino-5-n'-nitrocarbamimidamido pentanoic acid, boc arg no2 oh, boc-arg no-oh, pubchem12925, boc-l-arginine nitro, boc-l-arg no2 oh, boc-arg no2
PubChem CID 75141
SMILES CC(C)(C)OC(=O)NC(CCCN=C(N)N[N+](=O)[O-])C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6233914
View Documents
Thermo Scientific Chemicals
H6233914
25 g
Each for $164.36
Only null left
 
AAH6233922
View Documents
Thermo Scientific Chemicals
H6233922
100 g
Each for $508.36
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2188-18-3
319.318
OZSSOVRIEPAIMP-ZETCQYMHSA-N
75141
C11H21N5O6
MFCD00065556
boc-arg no2-oh, s-2-tert-butoxycarbonyl amino-5-3-nitroguanidino pentanoic acid, n2-tert-butoxycarbonyl-n5-n-nitrocarbamimidoyl-l-ornithine, 2s-2-tert-butoxycarbonyl amino-5-n'-nitrocarbamimidamido pentanoic acid, boc arg no2 oh, boc-arg no-oh, pubchem12925, boc-l-arginine nitro, boc-l-arg no2 oh, boc-arg no2
CC(C)(C)OC(=O)NC(CCCN=C(N)N[N+](=O)[O-])C(=O)O

Specifications

2188-18-3
MFCD00065556
boc-arg no2-oh, s-2-tert-butoxycarbonyl amino-5-3-nitroguanidino pentanoic acid, n2-tert-butoxycarbonyl-n5-n-nitrocarbamimidoyl-l-ornithine, 2s-2-tert-butoxycarbonyl amino-5-n'-nitrocarbamimidamido pentanoic acid, boc arg no2 oh, boc-arg no-oh, pubchem12925, boc-l-arginine nitro, boc-l-arg no2 oh, boc-arg no2
CC(C)(C)OC(=O)NC(CCCN=C(N)N[N+](=O)[O-])C(=O)O
319.318
319.31
Solid
C11H21N5O6
2015163
OZSSOVRIEPAIMP-ZETCQYMHSA-N
25 g
75141
98%
Nalpha-Boc-Nomega-nitro-L-arginine
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 218-580-0

TSCA : Yes

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only