CAS RN 2188-18-3
IUPAC Name:
(2S)-5-{[amino(nitroamino)methylidene]amino}-2-{[(tert-butoxy)carbonyl]amino}pentanoic acid
Synonyms:
(2S)-5-{[amino(nitroamino)methylidene]amino}-2-[(tert-butoxycarbonyl)amino]pentanoic acid
Molecular Weight (g/mol):
319.32
Molecular Formula:
C11H21N5O6
InChi Key:
OZSSOVRIEPAIMP-ZETCQYMHSA-N
SMILES:
CC(C)(C)OC(=O)N[C@@H](CCCN=C(N)N[N+]([O-])=O)C(O)=O
1
–
1
of
1
results
1
Promotions Available
Nalpha-Boc-N^w-nitro-L-arginine, 98%
Nalpha-Boc-N^w-nitro-L-arginine, 98%
CAS: 2188-18-3 Molecular Formula: C11H21N5O6 Molecular Weight (g/mol): 319.318 MDL Number: MFCD00065556 InChI Key: OZSSOVRIEPAIMP-ZETCQYMHSA-N Synonym: boc-arg no2-oh,s-2-tert-butoxycarbonyl amino-5-3-nitroguanidino pentanoic acid,n2-tert-butoxycarbonyl-n5-n-nitrocarbamimidoyl-l-ornithine,2s-2-tert-butoxycarbonyl amino-5-n'-nitrocarbamimidamido pentanoic acid,boc arg no2 oh,boc-arg no-oh,pubchem12925,boc-l-arginine nitro,boc-l-arg no2 oh,boc-arg no2 PubChem CID: 75141 SMILES: CC(C)(C)OC(=O)NC(CCCN=C(N)N[N+](=O)[O-])C(=O)O