missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-N^w-nitro-L-arginine, 98%

Catalog No. AAH6233922
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6233922 100 g
AAH6233914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6233922 Supplier Thermo Scientific Chemicals Supplier No. H6233922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2188-18-3 | C11H21N5O6 | 319.318 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2188-18-3
Molecular Formula C11H21N5O6
Molecular Weight (g/mol) 319.318
MDL Number MFCD00065556
InChI Key OZSSOVRIEPAIMP-ZETCQYMHSA-N
Synonym boc-arg no2-oh, s-2-tert-butoxycarbonyl amino-5-3-nitroguanidino pentanoic acid, n2-tert-butoxycarbonyl-n5-n-nitrocarbamimidoyl-l-ornithine, 2s-2-tert-butoxycarbonyl amino-5-n'-nitrocarbamimidamido pentanoic acid, boc arg no2 oh, boc-arg no-oh, pubchem12925, boc-l-arginine nitro, boc-l-arg no2 oh, boc-arg no2
PubChem CID 75141
SMILES CC(C)(C)OC(=O)NC(CCCN=C(N)N[N+](=O)[O-])C(=O)O

Specifications

Beilstein 2015163
Quantity 100 g
Formula Weight 319.31
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Nalpha-Boc-Nomega-nitro-L-arginine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-580-0
TSCA Yes
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only