missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-N^w-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-sulfonyl)-L-arginine, 96%

Catalog No. p-7058154
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6214803 1 g
AAH6214806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6214803 Supplier Thermo Scientific Chemicals Supplier No. H6214803
Only null left

CAS: 200124-22-7 | C24H38N4O7S | 526.649 g/mol

Nalpha-Boc-Nμ-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-sulfonyl)-L-arginine is used as a triflating reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nalpha-Boc-Nµ-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-sulfonyl)-L-arginine is used as a triflating reagent.

Solubility
Soluble in water or 1% acetic acid

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 200124-22-7
Molecular Formula C24H38N4O7S
Molecular Weight (g/mol) 526.649
MDL Number MFCD00236821
InChI Key CVFXPOKENLGCID-KRWDZBQOSA-N
Synonym boc-arg pbf-oh, s-2-tert-butoxycarbonyl amino-5-3-2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl sulfonyl guanidino pentanoic acid, n-boc-n'-2,2,4,6,7-pentamethyldihydrobenzofuran-5-sufonyl-l-arginine, ambotzbaa1067, boc-arg pbf-0h, n,a-boc-n, o-pbf-l-arginine, boc-arg pbf-oh tlc, o-2,2,4,6,7-pentamethyl-dihydrobenzofuran-5-sulfonyl-l-arginine, 2s-2-tert-butoxycarbonyl amino-5-n'-2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-ylsulfonyl carbamimidamido pentanoic acid, 2s-5-e-amino 2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-sulfonamido methylidene amino-2-tert-butoxycarbonyl amino pentanoic acid
PubChem CID 11341472
IUPAC Name (2S)-5-[[amino-[(2,2,4,6,7-pentamethyl-3H-1-benzofuran-5-yl)sulfonylamino]methylidene]amino]-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SMILES CC1=C2C(=C(C(=C1C)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OC(C)(C)C)N)C)CC(O2)(C)C

Specifications

Quantity 1 g
Solubility Information Soluble in water or 1% acetic acid
Formula Weight 526.65
Percent Purity 96%
Chemical Name or Material Nalpha-Boc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-sulfonyl)-L-arginine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only