missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-D-lysine, 98%

CAS: 106719-44-2 | C11H22N2O4 | 246.31 g/mol

$125.70 - $483.70

Chemical Identifiers

CAS 106719-44-2
Molecular Formula C11H22N2O4
Molecular Weight (g/mol) 246.31
MDL Number MFCD00076956
InChI Key DQUHYEDEGRNAFO-SVGMAFHSNA-N
Synonym boc-d-lys-oh, n-alpha-t-butoxycarbonyl-d-lysine, n2-boc-d-lysine, nalpha-tert-butoxycarbonyl-d-lysine, nalpha-tert-butoxycarbonyl-d-lysine, n-, a-boc-d-lysine, a-boc-d-lysine, d-lysine, n2-1,1-dimethylethoxy carbonyl, d-lysine, n2-1,1-dimethylethoxy carbonyl, n-tert-butoxycarbonyl-d-lysine
PubChem CID 7018770
IUPAC Name (2R)-6-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](CCCCN)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6296603
View Documents
Thermo Scientific Chemicals
H6296603
1 g
Each for $125.70
Only null left
 
AAH6296606
View Documents
Thermo Scientific Chemicals
H6296606
5 g
Each for $483.70
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

106719-44-2
246.31
DQUHYEDEGRNAFO-SVGMAFHSNA-N
7018770
CC(C)(C)OC(=O)N[C@H](CCCCN)C(O)=O
C11H22N2O4
MFCD00076956
boc-d-lys-oh, n-alpha-t-butoxycarbonyl-d-lysine, n2-boc-d-lysine, nalpha-tert-butoxycarbonyl-d-lysine, nalpha-tert-butoxycarbonyl-d-lysine, n-, a-boc-d-lysine, a-boc-d-lysine, d-lysine, n2-1,1-dimethylethoxy carbonyl, d-lysine, n2-1,1-dimethylethoxy carbonyl, n-tert-butoxycarbonyl-d-lysine
(2R)-6-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid

Specifications

106719-44-2
C11H22N2O4
1 g
DQUHYEDEGRNAFO-SVGMAFHSNA-N
CC(C)(C)OC(=O)N[C@H](CCCCN)C(O)=O
246.31
246.3
Nalpha-Boc-D-lysine
196°C to 200°C
MFCD00076956
boc-d-lys-oh, n-alpha-t-butoxycarbonyl-d-lysine, n2-boc-d-lysine, n2-boc-d-lysine, nalpha-boc-d-lysine, n-, n-, a-boc-d-lysine, d-lysine, n2-1,1-dimethylethoxy carbonyl, n-tert-butoxycarbonyl-d-lysine, n-tert-butoxycarbonyl-d-lysine
−22° (c=2 in Methanol)
(2R)-6-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
7018770
98%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only