missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-D-asparagine, 95%

CAS: 75647-01-7 | C9H16N2O5 | 232.236 g/mol

$75.19 - $293.04

Chemical Identifiers

CAS 75647-01-7
Molecular Formula C9H16N2O5
Molecular Weight (g/mol) 232.236
MDL Number MFCD00065558
InChI Key FYYSQDHBALBGHX-RXMQYKEDSA-N
Synonym boc-d-asparagine, boc-d-asn-oh, n-alpha-boc-d-asparagine, boc-l-asn-oh, nalpha-tert-butoxycarbonyl-d-asparagine, nalpha-boc-d-asparagine, n-boc-d-asparagine, n-alpha-t-butyloxycarbonyl-d-asparagine, d-asparagine, n2-1,1-dimethylethoxy carbonyl, 2r-2-tert-butoxycarbonyl amino-3-carbamoylpropanoic acid
PubChem CID 1623168
IUPAC Name (2R)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6112106
View Documents
Thermo Scientific Chemicals
H6112106
5 g
Each for $75.19
Only null left
 
AAH6112114
View Documents
Thermo Scientific Chemicals
H6112114
25 g
Each for $293.04
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

75647-01-7
232.236
FYYSQDHBALBGHX-RXMQYKEDSA-N
1623168
CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)O
C9H16N2O5
MFCD00065558
boc-d-asparagine, boc-d-asn-oh, n-alpha-boc-d-asparagine, boc-l-asn-oh, nalpha-tert-butoxycarbonyl-d-asparagine, nalpha-boc-d-asparagine, n-boc-d-asparagine, n-alpha-t-butyloxycarbonyl-d-asparagine, d-asparagine, n2-1,1-dimethylethoxy carbonyl, 2r-2-tert-butoxycarbonyl amino-3-carbamoylpropanoic acid
(2R)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid

Specifications

75647-01-7
C9H16N2O5
4843040
FYYSQDHBALBGHX-RXMQYKEDSA-N
(2R)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
232.236
232.23
Powder
White
MFCD00065558
boc-d-asparagine, boc-d-asn-oh, n-alpha-boc-d-asparagine, boc-l-asn-oh, nalpha-tert-butoxycarbonyl-d-asparagine, nalpha-boc-d-asparagine, n-boc-d-asparagine, n-alpha-t-butyloxycarbonyl-d-asparagine, d-asparagine, n2-1,1-dimethylethoxy carbonyl, 2r-2-tert-butoxycarbonyl amino-3-carbamoylpropanoic acid
CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)O
5 g
1623168
95%
Nalpha-Boc-D-asparagine
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only