missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-Boc-D-asparagine, 95%
CAS: 75647-01-7 | C9H16N2O5 | 232.236 g/mol
Supplier: Thermo Scientific Chemicals H6112114
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Nalpha-Boc-D-asparagine | |
| White | |
| MFCD00065558 | |
| boc-d-asparagine, boc-d-asn-oh, n-alpha-boc-d-asparagine, boc-l-asn-oh, nalpha-tert-butoxycarbonyl-d-asparagine, nalpha-boc-d-asparagine, n-boc-d-asparagine, n-alpha-t-butyloxycarbonyl-d-asparagine, d-asparagine, n2-1,1-dimethylethoxy carbonyl, 2r-2-tert-butoxycarbonyl amino-3-carbamoylpropanoic acid | |
| CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)O | |
| 25 g | |
| 1623168 | |
| 95% |
| 75647-01-7 | |
| C9H16N2O5 | |
| 4843040 | |
| FYYSQDHBALBGHX-RXMQYKEDSA-N | |
| (2R)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid | |
| 232.236 | |
| 232.23 | |
| Powder |
Chemical Identifiers
| 75647-01-7 | |
| 232.236 | |
| FYYSQDHBALBGHX-RXMQYKEDSA-N | |
| 1623168 | |
| CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)O |
| C9H16N2O5 | |
| MFCD00065558 | |
| boc-d-asparagine, boc-d-asn-oh, n-alpha-boc-d-asparagine, boc-l-asn-oh, nalpha-tert-butoxycarbonyl-d-asparagine, nalpha-boc-d-asparagine, n-boc-d-asparagine, n-alpha-t-butyloxycarbonyl-d-asparagine, d-asparagine, n2-1,1-dimethylethoxy carbonyl, 2r-2-tert-butoxycarbonyl amino-3-carbamoylpropanoic acid | |
| (2R)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only