missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Benzyloxycarbonyl-Ndelta-Boc-L-ornithine, 98%

CAS: 7733-29-1 | C18H26N2O6 | 366.414 g/mol

$175.12 - $683.18

Chemical Identifiers

CAS 7733-29-1
Molecular Formula C18H26N2O6
Molecular Weight (g/mol) 366.414
MDL Number MFCD00276410
InChI Key RWQCKACYKKSOKK-CQSZACIVSA-N
Synonym z-d-orn boc-oh, nalpha-z-ndelta-boc-d-ornithine, n-cbz-n'-boc-l-ornithine, na-z-nd-boc-d-ornithine, n2-benzyloxy carbonyl-n5-tert-butoxycarbonyl-d-ornithine, n∼2∼-benzyloxy carbonyl-n∼5∼-tert-butoxycarbonyl-d-ornithine, 2r-2-benzyloxy carbonyl amino-5-tert-butoxycarbonyl amino pentanoic acid, 2r-5-2-methylpropan-2-yl oxycarbonylamino-2-phenylmethoxycarbonylamino pentanoic acid, r-2-benzyloxycarbonyl amino-5-tert-butyloxycarbonyl amino pentanoic acid
PubChem CID 7350736
IUPAC Name (2R)-5-[(2-methylpropan-2-yl)oxycarbonylamino]-2-(phenylmethoxycarbonylamino)pentanoic acid
SMILES CC(C)(C)OC(=O)NCCCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6624706
View Documents
Thermo Scientific Chemicals
H6624706
5 g
Each for $175.12
Only null left
 
AAH6624714
View Documents
Thermo Scientific Chemicals
H6624714
25 g
Each for $683.18
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7733-29-1
366.414
RWQCKACYKKSOKK-CQSZACIVSA-N
7350736
CC(C)(C)OC(=O)NCCCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
C18H26N2O6
MFCD00276410
z-d-orn boc-oh, nalpha-z-ndelta-boc-d-ornithine, n-cbz-n'-boc-l-ornithine, na-z-nd-boc-d-ornithine, n2-benzyloxy carbonyl-n5-tert-butoxycarbonyl-d-ornithine, n∼2∼-benzyloxy carbonyl-n∼5∼-tert-butoxycarbonyl-d-ornithine, 2r-2-benzyloxy carbonyl amino-5-tert-butoxycarbonyl amino pentanoic acid, 2r-5-2-methylpropan-2-yl oxycarbonylamino-2-phenylmethoxycarbonylamino pentanoic acid, r-2-benzyloxycarbonyl amino-5-tert-butyloxycarbonyl amino pentanoic acid
(2R)-5-[(2-methylpropan-2-yl)oxycarbonylamino]-2-(phenylmethoxycarbonylamino)pentanoic acid

Specifications

97°C to 103°C
C18H26N2O6
3086976
RWQCKACYKKSOKK-CQSZACIVSA-N
(2R)-5-[(2-methylpropan-2-yl)oxycarbonylamino]-2-(phenylmethoxycarbonylamino)pentanoic acid
366.414
366.41
Solid
7733-29-1
MFCD00276410
z-d-orn boc-oh, nalpha-z-ndelta-boc-d-ornithine, n-cbz-n'-boc-l-ornithine, na-z-nd-boc-d-ornithine, n2-benzyloxy carbonyl-n5-tert-butoxycarbonyl-d-ornithine, n∼2∼-benzyloxy carbonyl-n∼5∼-tert-butoxycarbonyl-d-ornithine, 2r-2-benzyloxy carbonyl amino-5-tert-butoxycarbonyl amino pentanoic acid, 2r-5-2-methylpropan-2-yl oxycarbonylamino-2-phenylmethoxycarbonylamino pentanoic acid, r-2-benzyloxycarbonyl amino-5-tert-butyloxycarbonyl amino pentanoic acid
CC(C)(C)OC(=O)NCCCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
5 g
7350736
98%
Nalpha-Benzyloxycarbonyl-Ndelta-Boc-L-ornithine
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only