missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Benzyloxycarbonyl-L-2,3-diaminopropionic acid, 98%, Thermo Scientific Chemicals

Catalog No. p-7041411
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6202506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6202506 Supplier Thermo Scientific Chemicals Supplier No. H6202506

CAS: 35761-26-3 | Molecular Formula: C11H14N2O4 | Molecular Weight: 238.243 g/mol

Nalpha-Benzyloxycarbonyl-L-2,3-diaminopropionic acid is a important building block for the synthesis of peptides containing DAP residues, e.g. bleomycins, edeines, tuberactinomycins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nalpha-Benzyloxycarbonyl-L-2,3-diaminopropionic acid is a important building block for the synthesis of peptides containing DAP residues, e.g. bleomycins, edeines, tuberactinomycins.

Solubility
Soluble in dilute acid.

Notes
Hygroscopic. Store in cool, well ventilated area. Keep container tightly closed. Store away from water/moisture.

Chemical Identifiers

CAS 35761-26-3
Molecular Formula C11H14N2O4
Molecular Weight (g/mol) 238.243
MDL Number MFCD00237345
InChI Key FOXRXVSTFGNURG-VIFPVBQESA-N
Synonym z-dap-oh, cbz-beta-amino-l-alanine, 3-amino-2-n-cbz-l-alanine, s-3-amino-2-benzyloxy carbonyl amino propanoic acid, cbz-l-dap-oh, s-3-amino-2-benzyloxycarbonylamino propanoic acid, cbz-dap-oh, h-dap z-oh, n-alpha-cbz-l-2,3-diaminopropionic acid, z-dpr-oh
PubChem CID 2756313
IUPAC Name (2S)-3-amino-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CN)C(=O)O

Specifications

Melting Point ∼240°C (decomposition)
Beilstein 4486950
Solubility Information Soluble in dilute acid.
Quantity 5 g
Optical Rotation −39° (c-1 in 1M HCl)
Formula Weight 238.24
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material Nalpha-Benzyloxycarbonyl-L-2,3-diaminopropionic acid
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only