missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Benzoyl-L-arginine Ethyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. p-7118710
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B08535G 5 g
B085325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B08535G Supplier TCI America Supplier No. B08535G
Only null left

Chemical Identifiers

CAS 2645-08-1
Molecular Formula C15H23ClN4O3
Molecular Weight (g/mol) 342.82
MDL Number MFCD00012579
InChI Key HIXDELXKSSLIKB-UHFFFAOYNA-N
Synonym bz-arg-oet.hcl, n-alpha-benzoyl-l-arginine ethyl ester hydrochloride, ethyl n-benzoyl-l-argininate hydrochloride, baee, nalpha-benzoyl-l-arginine ethyl ester hydrochloride, l-baee, l-arginine, n2-benzoyl-, ethyl ester, monohydrochloride, n-benzoyl-l-arginine ethyl ester hydrochloride, ethyl n2-benzoyl-l-argininate monohydrochloride, ethyl 2s-5-carbamimidamido-2-phenylformamido pentanoate hydrochloride
PubChem CID 2723604
IUPAC Name ethyl (2S)-2-benzamido-5-(diaminomethylideneamino)pentanoate;hydrochloride
SMILES CCOC(=O)C(CCCN=C(N)N)NC(=O)C1=CC=CC=C1.Cl

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 5 g
Formula Weight 342.82
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-Benzoyl-L-arginine Ethyl Ester Hydrochloride