missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Benzoyl-L-argininamide Hydrochloride Monohydrate 98.0+%, TCI America™

Catalog No. B01021G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B01021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B01021G Supplier TCI America Supplier No. B01021G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4299-03-0
Molecular Formula C13H20ClN5O2
Molecular Weight (g/mol) 313.786
MDL Number MFCD00035003
InChI Key PYZACNCNNFUUDO-PPHPATTJSA-N
Synonym s-n-1-amino-5-guanidino-1-oxopentan-2-yl benzamide hydrochloride, n-alpha-benzoyl-l-argininamide hydrochloride, 2s-5-carbamimidamido-2-phenylformamido pentanamide hydrochloride, bz-arg-nh hcl, bz-arg-nh2 hcl, benzoyl-l-arginine amide hydrochloride, n-benzoyl-l-arginine amide hydrochloride, benzoyl-arginine amide monohydrochloride monihydrate, nalpha-benzoyl-l-argininamidehydrochloridemonohydrate, 2s-5-diaminomethylidene amino-2-phenylformamido pentanamide hydrochloride
PubChem CID 22871352
IUPAC Name N-[(2S)-1-amino-5-(diaminomethylideneamino)-1-oxopentan-2-yl]benzamide;hydrochloride
SMILES C1=CC=C(C=C1)C(=O)NC(CCCN=C(N)N)C(=O)N.Cl

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 313.79
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-Benzoyl-L-argininamide Hydrochloride Monohydrate