missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-tele-(triphenylmethyl)-D-histidine 97.0+%, TCI America™

Catalog No. p-7130082
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F11451G 1 g
F11455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F11451G Supplier TCI America Supplier No. F11451G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 135610-90-1
Molecular Formula C40H33N3O4
Molecular Weight (g/mol) 619.721
MDL Number MFCD00077061
InChI Key XXMYDXUIZKNHDT-DIPNUNPCSA-N
Synonym fmoc-d-his trt-oh, n-fmoc-n'-trityl-d-histidine, d-histidine, n-9h-fluoren-9-ylmethoxy carbonyl-1-triphenylmethyl, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1-triphenylmethyl imidazol-4-yl propanoic acid, pubchem18930, fmoc-d-his 1-trt-oh, fmoc-d-his-trt-oh, n-fmoc-1-trityl-d-histidine, fmoc-n-im-trityl-d-histidine, nalpha-fmoc-nim-trityl-d-histidine
PubChem CID 44828577
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(1-tritylimidazol-4-yl)propanoic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C=C(N=C4)CC(C(=O)O)NC(=O)OCC5C6=CC=CC=C6C7=CC=CC=C57

Specifications

Melting Point 135°C
Quantity 1 g
Formula Weight 619.72
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-tele-(triphenylmethyl)-D-histidine
TSCA No
Recommended Storage Refrigerator