missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-tert-butyl-L-threonine 98.0+%, TCI America™

Catalog No. F05051G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05051G 1 g
F05055G 5 g
F050525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F05051G Supplier TCI America Supplier No. F05051G
Only null left

Chemical Identifiers

CAS 71989-35-0
Molecular Formula C23H27NO5
Molecular Weight (g/mol) 397.47
MDL Number MFCD00077075
InChI Key LZOLWEQBVPVDPR-VLIAUNLRSA-N
Synonym fmoc-thr tbu-oh, fmoc-o-tert-butyl-l-threonine, fmoc-thr t-bu-oh, n-9-fluorenylmethoxycarbonyl-o-tert-butyl-l-threonine, fmoc-thr but, 2s,3r-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, n-9h-fluoren-9-yl methoxy carbonyl-o-tert-butyl-l-threonine, fmoc-thr but-oh, fmoc-o-tert.butyl-l-threonine, fmoc-thr otbu-oh
PubChem CID 6364643
IUPAC Name (2S,3R)-3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
SMILES C[C@@H](OC(C)(C)C)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

Specifications

Melting Point 133°C
Color White-Yellow
Quantity 1 g
Formula Weight 397.47
Percent Purity ≥98.0% (T,HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-tert-butyl-L-threonine