missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-biotinyl-L-lysine 95.0+%, TCI America™
$126.76 - $417.56
Chemical Identifiers
| CAS | 146987-10-2 |
|---|---|
| Molecular Formula | C31H38N4O6S |
| Molecular Weight (g/mol) | 594.73 |
| MDL Number | MFCD00270741 |
| InChI Key | OFIBQNGDYNGUEZ-CVKYCUBCNA-N |
| Synonym | fmoc-lys biotin-oh, fmoc-l-lys biotinyl-oh, fmoc-n-epsilon-biotinyl-l-lysine, nalpha-fmoc-nepsilon-biotinyl-l-lysine, nalpha-fmoc-nepsilon-biotinyl-l-lysine, fmoc biocytin, fmoc biocytin, fmoc-lys biotin, n, n, a-fmoc-biocytin |
| PubChem CID | 11039242 |
| IUPAC Name | (2S)-6-{5-[(3aS,4S,6aR)-2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl]pentanamido}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid |
| SMILES | [H][C@]12CS[C@@H](CCCCC(=O)NCCCC[C@H](NC(=O)OCC3C4=CC=CC=C4C4=CC=CC=C34)C(O)=O)[C@@]1([H])NC(=O)N2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
F1042200MG
|
TCI America
F1042200MG |
200 mg |
Each for $126.76
|
|
|||||
|
F10421G
|
TCI America
F10421G |
1 g |
Each for $417.56
|
|
|||||
Chemical Identifiers
| 146987-10-2 | |
| 594.73 | |
| OFIBQNGDYNGUEZ-CVKYCUBCNA-N | |
| 11039242 | |
| [H][C@]12CS[C@@H](CCCCC(=O)NCCCC[C@H](NC(=O)OCC3C4=CC=CC=C4C4=CC=CC=C34)C(O)=O)[C@@]1([H])NC(=O)N2 |
| C31H38N4O6S | |
| MFCD00270741 | |
| fmoc-lys biotin-oh, fmoc-l-lys biotinyl-oh, fmoc-n-epsilon-biotinyl-l-lysine, nalpha-fmoc-nepsilon-biotinyl-l-lysine, nalpha-fmoc-nepsilon-biotinyl-l-lysine, fmoc biocytin, fmoc biocytin, fmoc-lys biotin, n, n, a-fmoc-biocytin | |
| (2S)-6-{5-[(3aS,4S,6aR)-2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl]pentanamido}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid |
Specifications
| 183°C | |
| White | |
| MFCD00270741 | |
| OFIBQNGDYNGUEZ-CVKYCUBCNA-N | |
| (2S)-6-{5-[(3aS,4S,6aR)-2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl]pentanamido}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid | |
| 594.73 | |
| 594.73 | |
| Crystalline Powder |
| 146987-10-2 | |
| C31H38N4O6S | |
| fmoc-lys biotin-oh, fmoc-l-lys biotinyl-oh, fmoc-n-epsilon-biotinyl-l-lysine, nalpha-fmoc-nepsilon-biotinyl-l-lysine, nalpha-fmoc-nepsilon-biotinyl-l-lysine, fmoc biocytin, fmoc biocytin, fmoc-lys biotin, n, n, a-fmoc-biocytin | |
| [H][C@]12CS[C@@H](CCCCC(=O)NCCCC[C@H](NC(=O)OCC3C4=CC=CC=C4C4=CC=CC=C34)C(O)=O)[C@@]1([H])NC(=O)N2 | |
| 200 mg | |
| 11039242 | |
| ≥95.0% (HPLC,T) | |
| Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-biotinyl-L-lysine |
Safety and Handling
TSCA : No