missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-biotinyl-L-lysine 95.0+%, TCI America™
Supplier: TCI America F10421G
Specifications
| 183°C | |
| 146987-10-2 | |
| C31H38N4O6S | |
| fmoc-lys biotin-oh, fmoc-l-lys biotinyl-oh, fmoc-n-epsilon-biotinyl-l-lysine, fmoc-n-epsilon-biotinyl-l-lysine, nalpha-fmoc-nepsilon-biotinyl-l-lysine, fmoc biocytin, fmoc biocytin, fmoc-biocytin, n, a-fmoc-biocytin, a-fmoc-biocytin | |
| [H][C@]12CS[C@@H](CCCCC(=O)NCCCC[C@H](NC(=O)OCC3C4=CC=CC=C4C4=CC=CC=C34)C(O)=O)[C@@]1([H])NC(=O)N2 | |
| 1 g | |
| 11039242 | |
| ≥95.0% (HPLC,T) |
| Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-biotinyl-L-lysine | |
| White | |
| MFCD00270741 | |
| OFIBQNGDYNGUEZ-CVKYCUBCNA-N | |
| (2S)-6-{5-[(3aS,4S,6aR)-2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl]pentanamido}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid | |
| 594.73 | |
| 594.73 | |
| Crystalline Powder |
Chemical Identifiers
| 146987-10-2 | |
| 594.73 | |
| OFIBQNGDYNGUEZ-CVKYCUBCNA-N | |
| 11039242 | |
| [H][C@]12CS[C@@H](CCCCC(=O)NCCCC[C@H](NC(=O)OCC3C4=CC=CC=C4C4=CC=CC=C34)C(O)=O)[C@@]1([H])NC(=O)N2 |
| C31H38N4O6S | |
| MFCD00270741 | |
| fmoc-lys biotin-oh, fmoc-l-lys biotinyl-oh, fmoc-n-epsilon-biotinyl-l-lysine, fmoc-n-epsilon-biotinyl-l-lysine, nalpha-fmoc-nepsilon-biotinyl-l-lysine, fmoc biocytin, fmoc biocytin, fmoc-biocytin, n, a-fmoc-biocytin, a-fmoc-biocytin | |
| (2S)-6-{5-[(3aS,4S,6aR)-2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl]pentanamido}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid |
Safety and Handling
TSCA : No