missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-threonine tert-Butyl Ester 98.0+%, TCI America™
$369.08 - $1476.33
Chemical Identifiers
| CAS | 120791-76-6 |
|---|---|
| Molecular Formula | C23H27NO5 |
| Molecular Weight (g/mol) | 397.47 |
| MDL Number | MFCD08276378 |
| InChI Key | LMRQPSBMLDCJGN-UHFFFAOYNA-N |
| Synonym | Nalpha-Fmoc-L-threonine tert-Butyl Ester, Fmoc-Thr-OtBu |
| PubChem CID | 11773868 |
| IUPAC Name | tert-butyl 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-hydroxybutanoate |
| SMILES | CC(O)C(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)OC(C)(C)C |
Chemical Identifiers
| 120791-76-6 | |
| 397.47 | |
| LMRQPSBMLDCJGN-UHFFFAOYNA-N | |
| 11773868 | |
| CC(O)C(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)OC(C)(C)C |
| C23H27NO5 | |
| MFCD08276378 | |
| Nalpha-Fmoc-L-threonine tert-Butyl Ester, Fmoc-Thr-OtBu | |
| tert-butyl 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-hydroxybutanoate |
Specifications
| 100°C | |
| White | |
| MFCD08276378 | |
| LMRQPSBMLDCJGN-UHFFFAOYNA-N | |
| tert-butyl 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-hydroxybutanoate | |
| 397.47 | |
| 397.47 | |
| Crystalline Powder |
| 120791-76-6 | |
| C23H27NO5 | |
| Nalpha-Fmoc-L-threonine tert-Butyl Ester, Fmoc-Thr-OtBu | |
| CC(O)C(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)OC(C)(C)C | |
| 1 g | |
| 11773868 | |
| ≥98.0% (HPLC,N) | |
| Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-threonine tert-Butyl Ester |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator