CAS RN 120791-76-6
CAS RN 120791-76-6
IUPAC Name:
tert-butyl 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-hydroxybutanoate
Synonyms:
tert-butyl 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-hydroxybutanoate
Molecular Weight (g/mol):
397.47
Molecular Formula:
C23H27NO5
InChi Key:
LMRQPSBMLDCJGN-UHFFFAOYNA-N
SMILES:
CC(O)C(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)OC(C)(C)C
1
–
1
of
1
results
1
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-threonine tert-Butyl Ester 98.0+%, TCI America™
CAS: 120791-76-6 Molecular Formula: C23H27NO5 Molecular Weight (g/mol): 397.47 MDL Number: MFCD08276378 InChI Key: LMRQPSBMLDCJGN-UHFFFAOYNA-N Synonym: Nalpha-Fmoc-L-threonine tert-Butyl Ester, Fmoc-Thr-OtBu PubChem CID: 11773868 IUPAC Name: tert-butyl 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-hydroxybutanoate SMILES: CC(O)C(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)OC(C)(C)C