missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine tert-Butyl Ester 98.0+%, TCI America™

Catalog No. p-7129723
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05161G 1 g
F05165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05161G Supplier TCI America Supplier No. F05161G
Only null left

Chemical Identifiers

CAS 110797-35-8
Molecular Formula C22H25NO5
Molecular Weight (g/mol) 383.44
MDL Number MFCD01861331
InChI Key ZYOWIDHANLLHNO-UHFFFAOYNA-N
Synonym Nalpha-Fmoc-L-serine tert-Butyl Ester, Fmoc-Ser-OtBu
PubChem CID 11003502
IUPAC Name tert-butyl 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-hydroxypropanoate
SMILES CC(C)(C)OC(=O)C(CO)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 127°C
Color White
Quantity 1 g
Formula Weight 383.44
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine tert-Butyl Ester
TSCA No
Recommended Storage Refrigerator