missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-lysine Hydrochloride 98.0+%, TCI America™
$49.36 - $146.20
Chemical Identifiers
| CAS | 139262-23-0 |
|---|---|
| Molecular Formula | C21H25ClN2O4 |
| Molecular Weight (g/mol) | 404.89 |
| MDL Number | MFCD00190889 |
| InChI Key | MVMZFAIUUXYFGY-UPGKADEYNA-N |
| Synonym | fmoc-lys-oh.hcl, fmoc-lys-oh hydrochloride, fmoc-lys-oh hcl, nalpha-fmoc-l-lysine hydrochloride, fmoc-l-lysine hydrochloride, nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-lysine hydrochloride, l-lysine, n2-9h-fluoren-9-ylmethoxy carbonyl-, monohydrochloride, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-aminohexanoic acid hydrochloride, fmoc-l-lysine hcl, fmoc-lys-oh. hcl |
| PubChem CID | 11858928 |
| IUPAC Name | (2S)-6-amino-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid hydrochloride |
| SMILES | Cl.NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
Chemical Identifiers
| 139262-23-0 | |
| 404.89 | |
| MVMZFAIUUXYFGY-UPGKADEYNA-N | |
| 11858928 | |
| Cl.NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
| C21H25ClN2O4 | |
| MFCD00190889 | |
| fmoc-lys-oh.hcl, fmoc-lys-oh hydrochloride, fmoc-lys-oh hcl, nalpha-fmoc-l-lysine hydrochloride, fmoc-l-lysine hydrochloride, nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-lysine hydrochloride, l-lysine, n2-9h-fluoren-9-ylmethoxy carbonyl-, monohydrochloride, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-aminohexanoic acid hydrochloride, fmoc-l-lysine hcl, fmoc-lys-oh. hcl | |
| (2S)-6-amino-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid hydrochloride |
Specifications
| 139262-23-0 | |
| C21H25ClN2O4 | |
| fmoc-lys-oh.hcl, fmoc-lys-oh hydrochloride, fmoc-lys-oh hcl, nalpha-fmoc-l-lysine hydrochloride, fmoc-l-lysine hydrochloride, nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-lysine hydrochloride, l-lysine, n2-9h-fluoren-9-ylmethoxy carbonyl-, monohydrochloride, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-aminohexanoic acid hydrochloride, fmoc-l-lysine hcl, fmoc-lys-oh. hcl | |
| Cl.NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O | |
| 404.89 | |
| 404.89 | |
| Crystalline Powder |
| White | |
| MFCD00190889 | |
| MVMZFAIUUXYFGY-UPGKADEYNA-N | |
| (2S)-6-amino-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid hydrochloride | |
| 11858928 | |
| ≥98.0% (HPLC) | |
| Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-lysine Hydrochloride |
Safety and Handling
TSCA : No