missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-lysine Hydrochloride 98.0+%, TCI America™

Catalog No. F05861G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05861G 1 g
F05865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05861G Supplier TCI America Supplier No. F05861G

Chemical Identifiers

CAS 139262-23-0
Molecular Formula C21H25ClN2O4
Molecular Weight (g/mol) 404.89
MDL Number MFCD00190889
InChI Key MVMZFAIUUXYFGY-UPGKADEYNA-N
Synonym fmoc-lys-oh.hcl, fmoc-lys-oh hydrochloride, fmoc-lys-oh hcl, nalpha-fmoc-l-lysine hydrochloride, fmoc-l-lysine hydrochloride, nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-lysine hydrochloride, l-lysine, n2-9h-fluoren-9-ylmethoxy carbonyl-, monohydrochloride, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-aminohexanoic acid hydrochloride, fmoc-l-lysine hcl, fmoc-lys-oh. hcl
PubChem CID 11858928
IUPAC Name (2S)-6-amino-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid hydrochloride
SMILES Cl.NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Color White
Quantity 1 g
Formula Weight 404.89
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-lysine Hydrochloride