missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamine 98.0+%, TCI America™

Catalog No. p-7129606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03081G 1 g
F03085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03081G Supplier TCI America Supplier No. F03081G
Only null left

Chemical Identifiers

CAS 71989-20-3
Molecular Formula C20H20N2O5
Molecular Weight (g/mol) 368.39
MDL Number MFCD00037137
InChI Key IZKGGDFLLNVXNZ-UHFFFAOYNA-N
Synonym fmoc-gln-oh, nalpha-fmoc-l-glutamine, fmoc-l-gln-oh, fmoc-l-glutamine, n-fmoc-l-glutamine, fmoc-dap ac-oh, fmoc-gln, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-amino-5-oxopentanoic acid, 2s-4-carbamoyl-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, n alpha-fluorenylmethoxycarbonylglutamine
PubChem CID 2724775
IUPAC Name 4-carbamoyl-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
SMILES NC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Color White
Quantity 1 g
Formula Weight 368.39
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamine