CAS RN 71989-20-3
CAS RN 71989-20-3
IUPAC Name:
4-carbamoyl-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Synonyms:
4-carbamoyl-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanoic acid
Molecular Weight (g/mol):
368.39
Molecular Formula:
C20H20N2O5
InChi Key:
IZKGGDFLLNVXNZ-UHFFFAOYNA-N
SMILES:
NC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
1
–
1
of
1
results
1
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamine 98.0+%, TCI America™
CAS: 71989-20-3 Molecular Formula: C20H20N2O5 Molecular Weight (g/mol): 368.39 MDL Number: MFCD00037137 InChI Key: IZKGGDFLLNVXNZ-UHFFFAOYNA-N Synonym: fmoc-gln-oh,nalpha-fmoc-l-glutamine,fmoc-l-gln-oh,fmoc-l-glutamine,n-fmoc-l-glutamine,fmoc-dap ac-oh,fmoc-gln,s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-amino-5-oxopentanoic acid,2s-4-carbamoyl-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid,n alpha-fluorenylmethoxycarbonylglutamine PubChem CID: 2724775 IUPAC Name: 4-carbamoyl-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid SMILES: NC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O