missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-citrulline 98.0+%, TCI America™

Catalog No. F06261G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06261G 1 g
F06265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06261G Supplier TCI America Supplier No. F06261G

Chemical Identifiers

CAS 133174-15-9
Molecular Formula C21H23N3O5
Molecular Weight (g/mol) 397.431
MDL Number MFCD00151943
InChI Key NBMSMZSRTIOFOK-SFHVURJKSA-N
Synonym fmoc-cit-oh, fmoc-l-citrulline, nalpha-fmoc-l-citrulline, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-ureidopentanoic acid, l-ornithine, n5-aminocarbonyl-n2-9h-fluoren-9-ylmethoxy carbonyl, nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-citrulline, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-5-ureido-pentanoic acid, 2s-5-carbamoylamino-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, fmoc-citoh, pubchem19002
PubChem CID 2756127
IUPAC Name (2S)-5-(carbamoylamino)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCCNC(=O)N)C(=O)O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 397.43
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-citrulline