missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nalpha-4-Tosyl-L-arginine methyl ester hydrochloride, 98+%

CAS: 1784-03-8 | C14H22N4O4S·HCl | 378.88 g/mol

Supplier:  Thermo Scientific Chemicals 139200100

 View more versions of this product

Catalog No. AC139200100


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nα-4-Tosyl-L-arginine methyl ester hydrochloride
98.5
146°C to 149°C
Authentic
Glass bottle
H2NC(=NH)NH(CH2)3CH(NHSO2C6H4CH3)CO2CH3
10 g
tos-arg-ome.hcl, tame hydrochloride, tos-arg-ome hcl, nalpha-tosyl-l-arginine methyl ester hydrochloride, nalpha-p-tosyl-l-arginine methyl ester hydrochloride, nalpha-p-toluenesulfonyl-l-arginine methyl ester hydrochloride, tos-arg-omehcl, s-methyl 5-guanidino-2-4-methylphenylsulfonamido pentanoate hydrochloride, tosyl-l-arginine methyl ester hydrochloride, n-4-tosyl-l-arginine methyl ester hydrochloride
CC1=CC=C(C=C1)S(=O)(=O)NC(CCCN=C(N)N)C(=O)OC.Cl
378.88
378.88
Fine Crystalline Powder
1784-03-8
100.0
White
98+%
C14H22N4O4S·HCl
MFCD00012578
-14
JIQFFACVQXXHMY-YDALLXLXSA-N
methyl (2S)-5-(diaminomethylideneamino)-2-[(4-methylphenyl)sulfonylamino]pentanoate;hydrochloride
2723792
98+%

Chemical Identifiers

1784-03-8
378.88
JIQFFACVQXXHMY-YDALLXLXSA-N
2723792
CC1=CC=C(C=C1)S(=O)(=O)NC(CCCN=C(N)N)C(=O)OC.Cl
C14H22N4O4S·HCl
MFCD00012578
tos-arg-ome.hcl, tame hydrochloride, tos-arg-ome hcl, nalpha-tosyl-l-arginine methyl ester hydrochloride, nalpha-p-tosyl-l-arginine methyl ester hydrochloride, nalpha-p-toluenesulfonyl-l-arginine methyl ester hydrochloride, tos-arg-omehcl, s-methyl 5-guanidino-2-4-methylphenylsulfonamido pentanoate hydrochloride, tosyl-l-arginine methyl ester hydrochloride, n-4-tosyl-l-arginine methyl ester hydrochloride
methyl (2S)-5-(diaminomethylideneamino)-2-[(4-methylphenyl)sulfonylamino]pentanoate;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 217-235-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only