missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nalpha-4-Tosyl-L-arginine methyl ester hydrochloride, 98+%

Catalog No. p-3758840
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC139200050 5 g Glass bottle
AC139200100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC139200050 Supplier Thermo Scientific Chemicals Supplier No. 139200050
Only null left

CAS: 1784-03-8 | C14H22N4O4S·HCl | 378.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1784-03-8
Molecular Formula C14H22N4O4S·HCl
Molecular Weight (g/mol) 378.88
MDL Number MFCD00012578
InChI Key JIQFFACVQXXHMY-YDALLXLXSA-N
Synonym tos-arg-ome.hcl, tame hydrochloride, tos-arg-ome hcl, nalpha-tosyl-l-arginine methyl ester hydrochloride, nalpha-p-tosyl-l-arginine methyl ester hydrochloride, nalpha-p-toluenesulfonyl-l-arginine methyl ester hydrochloride, tos-arg-omehcl, s-methyl 5-guanidino-2-4-methylphenylsulfonamido pentanoate hydrochloride, tosyl-l-arginine methyl ester hydrochloride, n-4-tosyl-l-arginine methyl ester hydrochloride
PubChem CID 2723792
IUPAC Name methyl (2S)-5-(diaminomethylideneamino)-2-[(4-methylphenyl)sulfonylamino]pentanoate;hydrochloride
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC(CCCN=C(N)N)C(=O)OC.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 146°C to 149°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula H2NC(=NH)NH(CH2)3CH(NHSO2C6H4CH3)CO2CH3
Quantity 5 g
Specific Rotation -14
Formula Weight 378.88
Percent Purity 98+%
Physical Form Fine Crystalline Powder
Chemical Name or Material Nα-4-Tosyl-L-arginine methyl ester hydrochloride
Show More Show Less
EINECSNumber 217-235-1

RUO – Research Use Only