missing translation for 'onlineSavingsMsg'
Learn More

Nalidixic acid sodium salt

Nalixidic acid, CAS # 389-08-2, is a quinolone antibiotic that inhibits cell division. | CAS: 3374-05-8 | C12H11N2NaO3 | 254.22 g/mol

Supplier:  Thermo Scientific Chemicals J6355006

 View more versions of this product

Catalog No. AAJ6355006


Only null left
Explore available promotions

Description

Description

Nalidixic acid sodium salt is an antibacterial agent, which is inhibitor of bacterial DNA polymerase & avian myeloblastoma virus reverse transcriptase. The compound inhibits nucleic acid and protein synthesis in Saccharomyces cerevisiae.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Nalidixic acid is a monocarboxylic acid and quinoline antibiotic
• Nalixidic acid is a DNA synthesis inhibitor that acts by preventing DNA gyrase activity

Application

• Nalidixic acid is most effective against gram-negative bacteria, including Enterobacter species and other common resistant species
• It is suitable in laboratory applications for research of the mechanisms of gyrase activity and metabolic pathways related to DNA synthesis

Specifications

Specifications

Nalidixic acid sodium salt
>280°C
C12H11N2NaO3
5 g
nalidixic acid sodium salt, baktogram, nalidixic acid sodium, sodium nalidixate, chemiurin, dixilina, nalidixate sodium anhydrous, nalidixate sodium, nalidixate sodium anhydrous usan, 1-ethyl-1,4-dihydro-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid sodium salt
ROKRAUFZFDQWLE-UHFFFAOYSA-M
sodium;1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylate
3864541
Powder
5-8-3374
White to Yellow
MFCD00064376
3580062
Soluble at 50mg/ml in water
CCN1C=C(C(=O)C2=C1N=C(C=C2)C)C(=O)[O-].[Na+]
254.22
254.22

Chemical Identifiers

5-8-3374
254.22
ROKRAUFZFDQWLE-UHFFFAOYSA-M
3864541
CCN1C=C(C(=O)C2=C1N=C(C=C2)C)C(=O)[O-].[Na+]
C12H11N2NaO3
MFCD00064376
nalidixic acid sodium salt, baktogram, nalidixic acid sodium, sodium nalidixate, chemiurin, dixilina, nalidixate sodium anhydrous, nalidixate sodium, nalidixate sodium anhydrous usan, 1-ethyl-1,4-dihydro-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid sodium salt
sodium;1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

P261-P264b-P270-P272-P280g-P285-P301+P312-P302+P352-P304+P341-P330-P333+P313-P342+P311-P363-P501c

H302-H317-H334

EINECSNumber : 222-159-7

RTECSNumber : QN2885000

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only