missing translation for 'onlineSavingsMsg'
Learn More

Nalidixic acid sodium salt

Catalog No. p-7042726
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6355006 5 g
AAJ6355014 25 g
AAJ6355022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6355006 Supplier Thermo Scientific Chemicals Supplier No. J6355006
Only null left

Nalixidic acid, CAS # 389-08-2, is a quinolone antibiotic that inhibits cell division. | CAS: 3374-05-8 | C12H11N2NaO3 | 254.22 g/mol

Nalidixic acid sodium salt is an antibacterial agent, which is inhibitor of bacterial DNA polymerase & avian myeloblastoma virus reverse transcriptase. The compound inhibits nucleic acid and protein synthesis in Saccharomyces cerevisiae.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Nalidixic acid is a monocarboxylic acid and quinoline antibiotic
• Nalixidic acid is a DNA synthesis inhibitor that acts by preventing DNA gyrase activity

Application

• Nalidixic acid is most effective against gram-negative bacteria, including Enterobacter species and other common resistant species
• It is suitable in laboratory applications for research of the mechanisms of gyrase activity and metabolic pathways related to DNA synthesis

Chemical Identifiers

CAS 5-8-3374
Molecular Formula C12H11N2NaO3
Molecular Weight (g/mol) 254.22
MDL Number MFCD00064376
InChI Key ROKRAUFZFDQWLE-UHFFFAOYSA-M
Synonym nalidixic acid sodium salt, baktogram, nalidixic acid sodium, sodium nalidixate, chemiurin, dixilina, nalidixate sodium anhydrous, nalidixate sodium, nalidixate sodium anhydrous usan, 1-ethyl-1,4-dihydro-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid sodium salt
PubChem CID 3864541
IUPAC Name sodium;1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylate
SMILES CCN1C=C(C(=O)C2=C1N=C(C=C2)C)C(=O)[O-].[Na+]

Specifications

Melting Point >280°C
Color White to Yellow
Quantity 5 g
Beilstein 3580062
Solubility Information Soluble at 50mg/ml in water
Formula Weight 254.22
Physical Form Powder
Chemical Name or Material Nalidixic acid sodium salt
Hazard Category H302-H317-H334
Hazard Statement GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280g-P285-P301+P312-P302+P352-P304+P341-P330-P333+P313-P342+P311-P363-P501c
EINECSNumber 222-159-7
RTECSNumber QN2885000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only