Learn More
Nalidixic acid sodium salt, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 463550250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Nalidixic acid sodium salt | |
| C12H11N2NaO3 | |
| Solubility aq. soln.: 50 g/L (20°C). | |
| [Na+].CCN1C=C(C([O-])=O)C(=O)C2=CC=C(C)N=C12 | |
| 254.22 |
| 3374-05-8 | |
| 25 g | |
| ROKRAUFZFDQWLE-UHFFFAOYSA-M | |
| sodium 1-ethyl-7-methyl-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate | |
| 254.22 |
Chemical Identifiers
| 3374-05-8 | |
| 254.22 | |
| sodium 1-ethyl-7-methyl-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate |
| C12H11N2NaO3 | |
| ROKRAUFZFDQWLE-UHFFFAOYSA-M | |
| [Na+].CCN1C=C(C([O-])=O)C(=O)C2=CC=C(C)N=C12 |
Safety and Handling
GHS H Statement Harmful if swallowed.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
In case of inadequate ventilation wear respiratory protection.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Danger
EINECSNumber : 222-159-7
RUO â Research Use Only