CAS RN 3374-05-8
CAS RN 3374-05-8
IUPAC Name:
sodium 1-ethyl-7-methyl-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate
Synonyms:
sodium nalidixate
Molecular Weight (g/mol):
254.22
Molecular Formula:
C12H11N2NaO3
InChi Key:
ROKRAUFZFDQWLE-UHFFFAOYSA-M
SMILES:
[Na+].CCN1C=C(C([O-])=O)C(=O)C2=CC=C(C)N=C12
1
–
2
of
2
results
1
Nalidixic acid sodium salt, Thermo Scientific™
CAS: 3374-05-8 Molecular Formula: C12H11N2NaO3 Molecular Weight (g/mol): 254.22
2
Nalidixic acid sodium salt, Thermo Scientific™
CAS: 3374-05-8 Molecular Formula: C12H11N2NaO3 Molecular Weight (g/mol): 254.22 InChI Key: ROKRAUFZFDQWLE-UHFFFAOYSA-M IUPAC Name: sodium 1-ethyl-7-methyl-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate SMILES: [Na+].CCN1C=C(C([O-])=O)C(=O)C2=CC=C(C)N=C12