missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalidixic acid, ≥97.0% (TLC); ≥99.0% (T), MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Packaging | Bottomless Glass Bottle with Inserted Fused Cone |
| CAS | 389-08-2 |
| Physical Form | Neat |
| Linear Formula | C12H12N2O3 |
| MDL Number | MFCD00006884 |
| Melting Point | 227°C to 229°C |
| Synonym | 1,4-Dihydro-1-ethyl-7-methyl-1,8-naphthyridin-4-one-3-carboxylic acid; 1-Ethyl-1,4-dihydro-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 232.24 |
| Show More |