Learn More
Thermo Scientific™ Nalgene™ Wide-Mouth Opaque Amber HDPE Bottles with Closure: Bulk Pack

Store, ship and pack light-sensitive reagents in these wide-mouthed bottles that are leakproof when used with Nalgene closures.
Supplier: Thermo Scientific™ 3121060004
Description
Store, ship and pack light-sensitive reagents in Thermo ScientificTM NalgeneTM Wide-Mouth Opaque Amber HDPE Bottles with Closure.
- Opaque amber HDPE Bottles
- Opaque amber polypropylene closures
- Uniform walls resist splitting and puncturing
- Wide mouth allows easy filling and dispensing
- Bulk packed
- Leakproof†
Ordering Information:
Bottles and closures are packed in separate polybags within the same carton. † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Chemical Identifiers
| Opaque Amber | |
| HDPE | |
| Opaque Amber | |
| Wide | |
| Wide Mouth Bottle | |
| Screw (Bulk Packed, Unassembled) |
| Wide-Mouth Bottle | |
| Non-sterile | |
| Yes | |
| Round | |
| Polypropylene | |
| No |
Specifications
| 4 oz., 125 mL | |
| 4 oz. | |
| Wide-Mouth Bottle | |
| HDPE | |
| 500 | |
| Opaque Amber | |
| Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Classs VI. Product meets the requirements for light-resistant containers per USP latest edition. | |
| Wide | |
| 99 mm | |
| 3.89 in. | |
| 38 mm | |
| Screw (Bulk Packed, Unassembled) |
| 125 mL | |
| Opaque Amber | |
| 28 mm | |
| 500 | |
| Non-sterile | |
| 38 mm | |
| Yes | |
| Round | |
| Wide Mouth Bottle | |
| Polypropylene | |
| 50 mm | |
| No |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.