Learn More
Thermo Scientific™ Nalgene™ Wide-Mouth HDPE Bottles with Closure: Bulk Pack
Fill and dispense liquids using these wide-mouth HDPE bottles, which are leakproof when used with Nalgene closures.
$1466.62 - $6449.69
Specifications
Color Translucent
Description Wide-Mouth Bottle
Material HDPE
Sterility Non-sterile
Closure Color Natural
View More Specs
Products 6
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity  
03-313-79A
View Documents
Thermo Scientific™
3121040001
30 mL 28 mm
Case of 1000 Each for $6,352.69
Only null left
03-313-79B
View Documents
Thermo Scientific™
3121040002
60 mL 28 mm
Case of 1000 Each for $6,449.69
Only null left
03-313-79C
View Documents
Thermo Scientific™
3121040004
125 mL 38 mm
Case of 500 Each for $3,890.59
Only null left
03-313-79D
View Documents
Thermo Scientific™
3121040008
250 mL 43 mm
Case of 250 Each for $2,906.24
Only null left
03-313-79E
View Documents
Thermo Scientific™
3121040016
500 mL 53 mm
Case of 125 Each for $1,861.37
Only null left
03-313-79F
View Documents
Thermo Scientific™
3121040032
1 L 63 mm
Case of 50 Each for $1,466.62
Only null left
Description
Description
Fill and dispense liquid easily using these Thermo Scientific™ Nalgene™ Wide-Mouth HDPE Bottles with Closure. The uniform, natural high-density polyethylene walls resist splitting and puncturing.
Features:
  • Natural high-density polyethylene bottles with natural polypropylene closures
  • Excellent chemical resistance
  • Leakproof† when used with Nalgene closures
Ordering Information:
Bottles and closures are packed in separate polybags within the same carton.
Specifications
Specifications
Translucent
HDPE
Natural
Yes
Round
Polypropylene
No
Wide-Mouth Bottle
Non-sterile
Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Wide
Wide Mouth Bottle
Screw (Bulk Packed, Unassembled)
Videos
SDS
Documents
Documents
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.