missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Vented Unitary Right-to-Know LDPE Wash Bottles
GREENER_CHOICE

Catalog No. 0340920E
Change view
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Chemical Label:
Acetone
Distilled Water
Ethyl Alcohol
Isopropanol
Methanol
Sodium Hypochlorite
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Chemical Label
0340920E 500 mL Distilled Water
0340920C 500 mL Methanol
0340920D 500 mL Isopropanol
0340920B 500 mL Ethyl Alcohol
0340920F 500 mL Sodium Hypochlorite
0340920A 500 mL Acetone
0340921E 1000 mL Distilled Water
0340921A 1000 mL Acetone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 0340920E Supplier Thermo Scientific™ Supplier No. 24360505
Get a Water Bottle with Plastics Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Keep hazardous volatile liquids off the lab bench. Unitary wash bottles feature a unique vent system, and eliminate draw tube assembly and associated risk of contamination.

Keep hazardous volatile liquids off the lab bench with Thermo Scientific™ Nalgene™ Vented Unitary Right-To-Know LDPE Wash Bottles, featuring a unique vent system. Unitary bottle prevents contamination when filling the bottle; no draw tube assembly to pick up contamination off the lab bench. A strong, steady, no sputter stream of liquid is dispensed when the wash bottle is squeezed. Marked with four color, NFPA diamonds, GHS compliant hazard symbols, and right-to-know chemical information. Choose from seven chemical markings.

  • Bottle and tubulation molded in one piece to prevent chemical splash when removing closure and contamination when filling
  • Unique 38mm colored vented closure designed to prevent dripping
  • Sturdy polypropylene closures (HDPE on Sodium Hypochloride bottles)
  • Color-coded bar corresponds to closure, preventing cross-contamination
  • Prelabeled with ICS name and formula, as well as NFPA symbols for chemical handling, hazard, care and storage of six commonly-used chemicals
  • Leakproof†

Specifications

Capacity 16 oz., 500 mL
Capacity (Metric) 500 mL
Color Translucent
Description RTK Wash Bottle
Material LDPE
No. per Case 24
No. per Pack 4
Closure Color Natural
Unitary Yes
Closure Included Yes
Graduated No
Mouth Narrow
Chemical Label Distilled Water
Closure Material Polypropylene
Closure Type Screw Cap
Through the Closure Dispensing No
Autoclavable Not Autoclavable
Capacity (English) 16 oz.
Shape Cylindrical
Certifications/Compliance Meet OSHA requirements for workplace chemical container labeling.
Product Type Wash Bottle
Show More Show Less